missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-EDTA Buffer Solutions, Novus Biologicals™
Shop All Bio Techne Products
Click to view available options
pH:
8
9
Unit Size:
500mL
Description
- 10X EDTA buffer for Immunohistochemistry
- Dilute one part buffer with nine parts distilled water
- Unit size: 500mL
Recommended Storage
Store at room temperature
Specifications
Specifications
| Chemical Name or Material | Tris-EDTA Buffer |
| pH | 8 |
| Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction