missing translation for 'onlineSavingsMsg'
Learn More

NOX2 Rabbit, anti-Human, Clone: 1B17, Proteintech

Product Code. p-200050106 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054465 20 μL 20µL
18086855 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054465 Supplier Proteintech Supplier No. 828721RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The NOX2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human NOX2. This antibody recognizes human, mouse, rat antigen. The NOX2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen NOX2
Applications Western Blot, ELISA
Clone 1B17
Concentration 1000 μg/mL
Gene Symbols CGD, CGD91 phox, CYBB, Cytochrome b 245 heavy chain, Cytochrome b(558) subunit beta, Cytochrome b558 subunit beta, GP91 1, GP91 PHOX, GP91PHOX, NADPH oxidase 2, NOX2, p22 phagocyte B cytochrome, p91 PHOX
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1536
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.