missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPL Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93112-0.1ml
This item is not returnable.
View return policy
Description
NPL Polyclonal antibody specifically detects NPL in Mouse, Rat samples. It is validated for Western Blot
Specifications
| NPL | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C112, C1orf13MGC61869, chromosome 1 open reading frame 13, DHDPS1, dihydrodipicolinate synthetase homolog 1, EC 4.1.3.3, MGC149582, N-acetylneuraminate lyase, N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase), N-acetylneuraminate pyruvate-lyase, N-acetylneuraminic acid aldolase, NAL, NALase, NPL1, Sialate lyase, Sialate-pyruvate lyase, Sialic acid aldolase, Sialic acid lyase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human NPL (NP_110396.1). MAFPKKKLQGLVAATITPMTENGEINFSVIGQYVDYLVKEQGVKNIFVNGTTGEGLSLSVSERRQVAEEWVTKGKDKLDQVIIHVGALSLKESQELAQHAAEIGADGIAVIAPFFLKPWTKDILINFLKEVAAAAPALPFYYYHIPALTGVKIRAEELLDGILDKIPTFQGLKFSDTDLLDFGQCVDQNRQQQFAFLFGVDEQLLSALVMGATGAVGSTYNYLGKKTNQMLEAFEQKDFSLALNYQFCIQ | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 80896 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction