missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPS-1034, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
NPS-1034 is a dual inhibitor of AXL and MET with IC50s of 10.3 and 48 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | NPS-1034 |
| CAS | 1221713-92-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H23F2N5O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 34 mg/mL (61.65 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(C)N(C)N(C2=CC=C(F)C=C2)C1=O)NC3=CC=C(OC4=C5C(NC=C5C6=CC=CC=C6)=NC=C4)C(F)=C3 |
| Molecular Weight (g/mol) | 551.54 |
| Formula Weight | 551.54 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction