Learn More
Invitrogen™ NR5A2 Recombinant Rabbit Monoclonal Antibody (24GB390)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557136
 View more versions of this product
Product Code. 30275603
  • 146.00 € / 20µL
Description
NR5A2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Binds to the sequence element 5'-AACGACCGACCTTGAG-3' of the enhancer II of hepatitis B virus genes, a critical cis-element of their expression and regulation. May be responsible for the liver-specific activity of enhancer II, probably in combination with other hepatocyte transcription factors. Key regulator of cholesterol 7-alpha-hydroxylase gene (CYP7A) expression in liver. May also contribute to the regulation of pancreas-specific genes and play important roles in embryonic development.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NR5A2
Recombinant Monoclonal
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O00482, P45448, Q9QWM1
Nr5a2
A synthesized peptide derived from human NR5A2 (aa 450-524).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB390
Unconjugated
Nr5a2
Alpha-1-fetoprotein transcription factor; AU020803; B1-binding factor; b1-binding factor, hepatocyte transcription factor which activates enhancer II of hepatitis B virus; B1F; B1F2; CPF; CYP7A promoter-binding factor; D1Ertd308e; fetoprotein transcription factor; fetoprotein-alpha 1 (AFP) transcription factor; Ftf; FTF/LRH-1; FTZ F1; FTZ F1beta; FTZ-F1; FTZ-F1 beta; FTZ-F1 beta2 protein; FTZ-F1beta; hB1F; hB1F 2; hB1F-2; Hepatocytic transcription factor; hepatocytic transcription factor hB1F-3; I79_021747; live; liver nuclear receptor homolog-1 variant 2; liver receptor homolog 1; liver receptor homolog-1; Lrh1; LRH-1; Nr5a2; Nuclear receptor subfamily 5 group A member 2; nuclear receptor subfamily 5, group A, member 2; OR2.0; orphan nuclear receptor; SF-2; steroidogenic factor 2; UF2-H3B
Rabbit
Affinity chromatography
RUO
2494, 26424, 60349
-20°C