missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRAS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99926-50ul
This item is not returnable.
View return policy
Description
NRAS Polyclonal antibody specifically detects NRAS in Human samples. It is validated for ELISA
Specifications
| NRAS | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| ALPS4, GTPase NRas, HRAS1, neuroblastoma RAS viral (v-ras) oncogene homolog, N-ras, N-ras protein part 4, NRAS1, NS6, Transforming protein N-Ras, v-ras neuroblastoma RAS viral oncogene homolog | |
| Produced in rabbits immunized with purified, recombinant Human NRAS (Accession#: P01111; Met1-Cys186) | |
| 50 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, mTOR Pathway | |
| 4893 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction