missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99226-50ul
This item is not returnable.
View return policy
Description
NRBP2 Polyclonal antibody specifically detects NRBP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NRBP2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| DKFZp434P086, MGC138699, nuclear receptor binding protein 2, nuclear receptor-binding protein 2, pp9320, Transformation-related gene 16 protein, transformation-related protein 16, TRG16, TRG-16 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NRBP2. | |
| 50 μg | |
| Protein Kinase | |
| 340371 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction