missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nrf2 Polyclonal Antibody

Product Code. 13411847
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
13411847 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 13411847

Brand: Invitrogen™ PA527882

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: C8D30, 3xFlag-human NFE2L2-transfected 293T, MDA-MB-231 nuclear extract, MDA-MB-231 nuclear extract (30 μM tBHQ, 4hr), HepG2 (10 μM MG132 treatment for 15 hr). Predicted reactivity: Rhesus Monkey (99%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nrf2
Applications ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene NFE2L2
Gene Accession No. O54968, Q16236, Q60795
Gene Alias AI194320; E4TF1-60; E4TF1A; HEBP1; Nfe2l2; NF-E2-related factor 2; NFE2-related factor 2; NF-E2-related factor-2; NFT2; NRF2; NRF2A; nuclear factor (erythroid-derived 2)-like 2; nuclear factor erythroid 2-like 2; nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2 like 2; nuclear factor, erythroid 2-like 2; nuclear factor, erythroid derived 2, like 2; Unknown (protein for IMAGE:7945830)
Gene Symbols NFE2L2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NRF2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18024, 4780, 83619
Target Species Human, Mouse, Rat, Avian, Reptile, Zebrafish, Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.