missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRGN Recombinant Rabbit Monoclonal Antibody (4D4V2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558046
This item is not returnable.
View return policy
Description
Immunogen sequence: DCCTENACSK PDDDILDIPL DDPGANAAAA KIQASFRGHM ARKKIKSGER GRKGPGPGGP GGAGVARGGA GGGPSGD.
Neurogranin is the human homolog of the neuron-specific rat RC3/neurogranin gene. This gene encodes a postsynaptic protein kinase substrate that binds calmodulin in the absence of calcium. The NRGN gene contains four exons and three introns. The exons 1 and 2 encode the protein and exons 3 and 4 contain untranslated sequences. It is suggested that the NRGN is a direct target for thyroid hormone in human brain, and that control of expression of this gene could underlay many of the consequences of hypothyroidism on mental states during development as well as in adult subjects.
Specifications
| NRGN | |
| Recombinant Monoclonal | |
| 1.35 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P60761, Q04940 | |
| NRGN | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4D4V2 | |
| Unconjugated | |
| NRGN | |
| 0710001B06Rik; AI838505; B-50 immunoreactive C-kinase substrate; BICKS; calmodulin-binding protein; hng; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); ng; NG/RC3; NRGN; p17; postsynaptic spine 1; protein kinase C substrate; protein kinase C substrate 7.5 kDa protein; protein kinase C substrate, RC3; Pss1; R75334; RC3; similar to neurogranin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 64011, 64356 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction