missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 1643

Description
Human ether-a-go-go related gene (hERG) KV11.1 channel activator (EC50 = 10.5 μM). Exhibits different molecular mechanisms of action at KV11.1 (hERG1) and KV11.2 (hERG2) channels. Displays antiarrhythmic activity in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 380.24 |
| Purity | >98% |
| Molecular Weight (g/mol) | 380.246 |
| Chemical Name or Material | NS 1643 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?