CAS RN 448895-37-2
CAS RN 448895-37-2
IUPAC Name:
1,3-bis[2-hydroxy-5-(trifluoromethyl)phenyl]urea
Synonyms:
1,3-bis[2-hydroxy-5-(trifluoromethyl)phenyl]urea
Molecular Weight (g/mol):
380.25
Molecular Formula:
C15H10F6N2O3
InChi Key:
NJFVQMRYJZHGME-UHFFFAOYSA-N
SMILES:
OC1=C(NC(=O)NC2=C(O)C=CC(=C2)C(F)(F)F)C=C(C=C1)C(F)(F)F
1
–
2
of
2
results
1
2
NS1643, MedChemExpress
MedChemExpress NS1643 is a partial agonist of human ether-a-go-go-related gene (hERG) K(+) channels with an EC50 of 10.5 μM.