missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 1643
GREENER_CHOICE

Product Code. 18095712
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18095712 50 mg 50mg
18065802 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095712 Supplier Tocris Bioscience™ Supplier No. 3062/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KV11.1 (hERG) channel activator; antiarrhythmic

Human ether-a-go-go related gene (hERG) KV11.1 channel activator (EC50 = 10.5 μM). Exhibits different molecular mechanisms of action at KV11.1 (hERG1) and KV11.2 (hERG2) channels. Displays antiarrhythmic activity in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448895-37-2
Molecular Formula C15H10F6N2O3

Specifications

Quantity 50 mg
Formula Weight 380.24
Purity >98%
Molecular Weight (g/mol) 380.246
Chemical Name or Material NS 1643
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.