missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NS-2028, MedChemExpress
Description
NS-2028 inhibits soluble Guanylyl Cyclase activity in homogenates of mouse cerebellum and neuronal NO synthase with IC50 values of 17 nM and 20 nM. NS-2028 inhibits 3-morpholino-sydnonimine (SIN-1)-elicited formation of cyclic GMP in human cultured umbilical vein endothelial cells with an IC50 of 30 nM. NS-2028 is commonly used in the research of nitric oxide signaling pathways, it inhibits NO-dependent relaxant responses in non-vascular smooth muscle completely (1 μM). NS-2028 reduces vascular endothelial growth factor-induced angiogenesis and permeability.
Specifications
Specifications
| Chemical Name or Material | NS-2028 |
| CAS | 204326-43-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C9H5BrN2O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 250 mg/mL (929.20 mM; Need ultrasonic) |
| SMILES | O=C1ON=C2COC3=CC=C(Br)C=C3N21 |
| Molecular Weight (g/mol) | 269.05 |
| Formula Weight | 269.05 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?