missing translation for 'onlineSavingsMsg'
Learn More

NS1643, MedChemExpress

Product Code. 16497208
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497208 10 mg 10mg
16497186 100 mg 100mg
16493227 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497208 Supplier Medchem Express Supplier No. HY16916

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NS1643 is a partial agonist of human ether-a-go-go-related gene (hERG) K(+) channels with an EC50 of 10.5 μM.

NS1643 is a partial agonist of human ether-a-go-go-related gene (hERG) K(+) channels with an EC50 of 10.5 μM.

Specifications

Chemical Name or Material NS1643
CAS 448895-37-2
Color White
Purity Grade Notes Research
Molecular Formula C15H10F6N2O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (262.99 mM)
SMILES O=C(NC1=C(O)C=CC(C(F)(F)F)=C1)NC2=C(O)C=CC(C(F)(F)F)=C2
Molecular Weight (g/mol) 380.24
Formula Weight 380.24
Percent Purity 95.03%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.