missing translation for 'onlineSavingsMsg'
Learn More

NSC 87877, Tocris Bioscience™

Product Code. 30001301
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001301 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001301 Supplier Tocris Bioscience Supplier No. 2613/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent inhibitor of shp2 and shp1 PTP

Potent inhibitor of shp2 and shp1 protein tyrosine phosphatases (PTP) (IC50 values are 0.318, 0.355, 1.691, 7.745, 65.617, 84.473 and 150.930 μM for shp2, shp1, PTP1B, HePTP, DEP1, CD45 and LAR respectively). Inhibits EGF-induced Erk1/2 activation in HEK293 cells and significantly reduces MDA-MB-468 cell viability/proliferation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 56990-57-9
Molecular Formula C19H13N3O7S2
Quantity 50 mg
Synonym 8-hydroxy-7-6-sulfo-naphthalen-2-ylazo-quinoline-5-sulfonic acid, 8-hydroxy-7-6-sulfo-2-naphthyl azo-5-quinolinesulfonic acid, ncistruc1_001562, ncistruc2_001738, 8-hydroxy-7-6-sulfonaphthalen-2-yl diazenyl-quinoline-5-sulfonic acid, 8-hydroxy-7-6-sulfonaphthalen-2-yl diazenyl quinoline-5-sulfonic acid, 8-hydroxy-7-e-6-sulfo-2-naphthyl azo quinoline-5-sulfonic acid, 7e-8-oxo-7-6-sulfonaphthalen-2-yl hydrazinylidene quinoline-5-sulfonic acid
InChI Key BIQBDHSYQMHHPU-CJLVFECKSA-N
SMILES C1=CC2=C(C(=O)C(=NNC3=CC4=C(C=C3)C=C(C=C4)S(=O)(=O)O)C=C2S(=O)(=O)O)N=C1
IUPAC Name (7E)-8-oxo-7-[(6-sulfonaphthalen-2-yl)hydrazinylidene]quinoline-5-sulfonic acid
Molecular Weight (g/mol) 459.447
PubChem CID 5459322
Formula Weight 459.45
Percent Purity >98%
Chemical Name or Material NSC 87877
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.