missing translation for 'onlineSavingsMsg'
Learn More

NSC16168, MedChemExpress

Product Code. 16479676
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479676 50 mg 50mg
16454416 100 mg 100mg
16445857 5 mg 5mg
16413128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16479676 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY100690
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.

NSC16168 is a specific inhibitor of ERCC1-XPF, with an IC50 value of 0.42 μM. NSC16168 inhibits DNA repair and potentiates CDDP efficacy in cancer.

Specifications

Chemical Name or Material NSC16168
CAS 6837-93-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C17H15NO9S3
Quantity 50 mg
Solubility Information DMSO : 31.25 mg/mL (66.00 mM; Need ultrasonic)
SMILES NC1=C2C(C=C(S(=O)(O)=O)C=C2OS(C3=CC=C(C)C=C3)(=O)=O)=CC(S(O)(=O)=O)=C1
Molecular Weight (g/mol) 473.5
Formula Weight 473.5
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.