Learn More
Invitrogen™ NSDHL Recombinant Rabbit Monoclonal Antibody (24GB510)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557524
 View more versions of this product
Product Code. 30275147
  • 564.00 € / 100µL
Description
NSDHL Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NSDHL
Recombinant Monoclonal
Unconjugated
Nsdhl
AI747449; bare patches; Bpa; H105E3; NAD(P) dependent steroid dehydrogenase-like; NAD(P)-dependent steroid dehydrogenase-like protein; Nsdhl; Protein H105e3; SDR31E1; short chain dehydrogenase/reductase family 31E, member 1; sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; Str; striated; XAP104
Rabbit
Affinity chromatography
RUO
18194, 309262, 50814
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB510
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q15738, Q5PPL3, Q9R1J0
Nsdhl
A synthesized peptide derived from human NSDHL (aa 100-150).
100 μL
Primary
Human, Mouse, Rat