missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 572.00 €
Specifications
| Antigen | NSUN3 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18430041
|
Novus Biologicals
NBP1-83595-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18245917
|
Novus Biologicals
NBP1-83595 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NSUN3 Polyclonal specifically detects NSUN3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NSUN3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 63899 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PGGKSIALLQCACPGYLHCNEYDSLRLRWLRQTLESFIPQPLINVIKVSELDGRKMGDAQPEMFDKVLVDAPCSNDRSWLFSSDSQKASCRISQR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 2.1.1, EC 2.1.1.-, FLJ22109, FLJ22609, MST077, NOL1/NOP2/Sun domain family 3, NOL1/NOP2/Sun domain family member 3, NOL1/NOP2/Sun domain family, member 3, NOP2/Sun domain family, member 3, putative methyltransferase NSUN3 | |
| NSUN3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title