missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NTB Binding Buffer

Brand:  Macherey-Nagel™ 740595.150

Code : LE

Additional Details : CAS Number : 50-01-1 Weight : 0.28800kg Transport : UN number : 1993 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 11952282

  • 43.75 € / 150mL
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The NucleoSpin™ Extract II procedure is the easiest way to purify DNA fragments from agarose gels as well as for direct purification of PCR products. The purification procedure from enzymatic reactions (eg. PCR) allows fast and easy removal of enzymes, nucleotides, salts and other impurities. The NucleoSpin™ Extract II columns provide convenient performance for PCR clean-up. After addition of binding buffer NT the mixture is applied onto the silica membrane. Contaminations are removed by a simple washing step with ethanolic buffer NT3.Pure DNA is finally eluted under low ionic strength conditions with slightly alkaline buffer NE (5mM Tris-HCl, pH8.5). For DNA extraction from agarose gels the agarose gel slice is incubated with high-salt buffer and applied to a NucleoSpin™ Extract II column. After centrifugation and subsequent washing the DNA can be eluted under low ionic strength conditions with slightly alkaline buffer NE (5mM Tris-HCl, pH8.5). Kit components: NucleoSpin™ Extract II columns, collecting tubes 2mL, buffers.
  • Silica membrane technology
  • Yield: 70% to 95%
  • Sample material: <400μL PCR reaction mixture; <400mg TAE/TBE agarose gel
  • High recovery of fragments 65bp to 10kb
  • Elution volume: 15μL to 50μL
  • Binding capacity: 25μL
  • Preparation time: 10min/6 preps
  • Format: mini spin column For direct purification of PCR* products and extraction of DNA from agarose gels.
Specifications

Specifications

150 mL
For Clean-up of SDS-Containing Samples
Binding Buffer NTB
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • buffer NTB

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash with water thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride, propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers