missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTH1 Antibody (S08-8E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19722-100ul
This item is not returnable.
View return policy
Description
NTH1 Monoclonal antibody specifically detects NTH1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NTH1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-8E9 | |
| Unconjugated | |
| EC 4.2.99.18, endonuclease III-like protein 1, nth endonuclease III-like 1 (E. coli), OCTS3NTH1nth (E.coli endonuclease III)-like 1 | |
| Recombinant protein of human NTH1 (Uniprot # P78549) | |
| 100 μg | |
| Base Excision Repair, Cancer, Chromatin Research, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4913 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction