missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUBP1 Monoclonal Antibody (2B11)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA519412
This item is not returnable.
View return policy
Description
Peptide Sequence: PIIGVVENMS GFICPKCKKE SQIFPPTTGG AELMCQDLEV PLLGRVPLDP LIGKNCDKGQ SFFIDAPDSP ATLAYRSIIQ RIQEFCNLHQ SKEENLISS.
Component of the cytosolic iron-sulfur (Fe/S) protein assembly (CIA) machinery (PubMed:18573874). Required for maturation of extramitochondrial Fe-S proteins (PubMed:18573874). The NUBP1-NUBP2 heterotetramer forms a Fe-S scaffold complex, mediating the de novo assembly of an Fe-S cluster and its transfer to target apoproteins (PubMed:18573874). Implicated in the regulation of centrosome duplication. Negatively regulates cilium formation and structure.
Specifications
| NUBP1 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P53384 | |
| NUBP1 | |
| NUBP1 (NP_002475.1, 222 a.a. ~ 320 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2B11 | |
| Unconjugated | |
| NUBP1 | |
| cytosolic Fe-S cluster assembly factor NUBP1; cytosolic Fe-S cluster assembly factor NUBP1 {ECO:0000255; EMBL:AAH88221.1}; HAMAP-Rule:MF_03038}; NBP; NBP 1; NBP 1 {ECO:0000255; NBP1; NBP35; NUBP1; nubp1 {ECO:0000312; nucleotide binding protein (e.coli MinD like); nucleotide binding protein 1; nucleotide binding protein 1 (E.coli MinD like); nucleotide binding protein 1 (MinD homolog, E. coli); nucleotide-binding protein 1; nucleotide-binding protein 1 {ECO:0000255 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4682 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction