missing translation for 'onlineSavingsMsg'
Learn More

Nuclear Antigen Antibody (235-1), PE, Novus Biologicals™

Mouse Monoclonal Antibody

595.00 € - 643.00 €

Specifications

Antigen Nuclear Antigen
Clone 235-1
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18450812
View Documents
Novus Biologicals
NBP2-34525PE-100ul
100 μL
643.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18498841
View Documents
Novus Biologicals
NBP2-34525PE
0.1 mL
595.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Porcine, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF
Specifications

Specifications

Nuclear Antigen
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready
Monoclonal
Purified
RUO
PBS
Nuclei of human myeloid leukemia biopsy cells
Primary
Store at 4C in the dark.
235-1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry (Intracellular Staining), CyTOF
PE
Mouse
Human, Pig, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative)
Nuclei, Nucleus
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.