Learn More
Invitrogen™ Nuclear Antigen Monoclonal Antibody (235-1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533098
Description
This antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells, including neurons and embryonic stem cells, with no reactivity to rat or mouse cells. This clone stains nuclei of all human cell types giving a diffuse nuclear staining pattern; chromosomes are negatively stained in metaphase cells. With immunoprecipitation, the antibody reacts with a ~80 kDa and ~70 kDa band. The antibody works on acetone-fixed and paraformaldehyde-fixed cells. It is suggested to permeabilize the cells with 0.1-0.4% Triton X-100. The antibody also works for immunohistochemistry on paraformaldehyde fixed cryostat sections.
Specifications
| Nuclear Antigen | |
| Monoclonal | |
| 1 mg/mL | |
| Tissue culture supernatant with no preservative | |
| Mouse | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| 235-1 | |
| Unconjugated | |
| Histone H1; HNA; HuNu; Nuclei; Nucleus | |
| Human cell homogenate. | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.