missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Factor Erythroid Derived 2 Antibody (SR1755), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22631-100ul
This item is not returnable.
View return policy
Description
Nuclear Factor Erythroid Derived 2 Monoclonal antibody specifically detects Nuclear Factor Erythroid Derived 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Serum
Specifications
| Nuclear Factor Erythroid Derived 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 4778 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1755 | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Serum 1:50 | |
| Leucine zipper protein NF-E2, NF-E2, nuclear factor (erythroid-derived 2), 45kD, nuclear factor (erythroid-derived 2), 45kDa, Nuclear factor, erythroid-derived 2 45 kDa subunit, p45, p45 NF-E2, transcription factor NF-E2 45 kDa subunit | |
| A synthesized peptide derived from human Nuclear Factor Erythroid Derived 2 (Uniprot #: Q16621) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction