missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nuclear Matrix Protein p84 Recombinant Rabbit Monoclonal Antibody (24GB2205)

Rabbit Recombinant Monoclonal Antibody
198.00 € - 564.00 €
Specifications
| Antigen | Nuclear Matrix Protein p84 |
|---|---|
| Clone | 24GB2205 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276526
![]() |
Invitrogen™
MA557772 |
100 μL |
564.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274872
|
Invitrogen™
MA557773 |
20 μL |
198.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nuclear Matrix Protein p84 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).Specifications
| Nuclear Matrix Protein p84 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Thoc1 | |
| 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1 | |
| Thoc1 | |
| Primary | |
| Affinity Chromatography |
| 24GB2205 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P59924, Q8R3N6, Q96FV9 | |
| 225160, 291797, 9984 | |
| A synthesized peptide derived from human Nuclear Matrix Protein p84 (aa 350-400). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title