missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nucleodur™ C18 Gravity HPLC CC Guard Columns

Brand:  Macherey-Nagel™ 761125.30

Code : 16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10011872

  • 213.00 € / Pack of 3

Estimated Shipment: 05-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

18%
5 μm
110Å
3 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
Yes
1 to 11
8 mm
CC Guard Column
NUCLEODUR™ C18 Gravity
Acetonitrile/Water
Octadecyl
Separations Pharmaceuticals, Derivatised Amino Acids, Drugs
Spherical
L1
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers