missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleolin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00 € - 456.00 €
Specifications
| Antigen | Nucleolin |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453021
|
Novus Biologicals
NBP1-88271 |
0.1 mL |
456.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18461771
|
Novus Biologicals
NBP1-88271-25ul |
25 μL |
302.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nucleolin Polyclonal antibody specifically detects Nucleolin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Nucleolin | |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol | |
| 4691 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VLSNLSYSATEETLQEVFEKATFIKVPQNQNGKSKGYAFIEFASFEDAKEALNSCNKREIEGRAIRLELQGPRGSPNARSQPSKTLFVKGLSEDTTEETLKESFDGSVRARIVTDRETGSSKGFGFVDFNSEEDA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title