missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97160JF525
This item is not returnable.
View return policy
Description
Nudel Polyclonal antibody specifically detects Nudel in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Nudel | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 525 | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| Produced in rabbits immunized with E. coli-derived Human Nudel fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 81565 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction