missing translation for 'onlineSavingsMsg'
Learn More

NUDT21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16866633 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16866633 20 μL 20µL
16856633 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16866633 Supplier Proteintech Supplier No. 103221AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NUDT21 is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitment of other processing factors. The protein encoded by this gene is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitment of other processing factors. This gene encodes the 25kD subunit of the protein complex, which is composed of four polypeptides.

Specifications

Antigen NUDT21
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NUDT21
Gene Accession No. O43809, Q4KM65, Q9CQF3
Gene Alias CFIM25, CPSF 25 kDa subunit, CPSF25, CPSF5, Nudix motif 21, NUDT21
Gene Symbols Nudt21
Host Species Rabbit
Immunogen NUDT21 Fusion Protein Ag0393
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11051, 291877, 68219
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.