Learn More
NUDT9 Antibody (S07-9C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19723-50ul
 View more versions of this product
Product Code. 18626593
  • 421.00 € / 50µL
Description
NUDT9 Monoclonal antibody specifically detects NUDT9 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NUDT9
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified
Western Blot, Immunofluorescence
S07-9C3
Unconjugated
ADP-ribose diphosphatase, ADP-ribose phosphohydrolase, ADP-ribose pyrosphosphatase NUDT9, ADPR-PPase, EC 3.6.1.13, MGC3037, mitochondrial, nucleoside diphosphate linked moiety X-type motif 9, nudix (nucleoside diphosphate linked moiety X)-type motif 9, Nudix motif 9, NUDT10
A synthetic peptide of human NUDT9 (Uniprot # Q9BW91)
50 μg
Signal Transduction
53343
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.