missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuf2 Antibody (S07-9E2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19724-50ul
This item is not returnable.
View return policy
Description
Nuf2 Monoclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western Blot
Specifications
| Nuf2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S07-9E2 | |
| Unconjugated | |
| cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
| A synthetic peptide of human Nuf2 (Uniprot # Q9BZD4) | |
| 50 μg | |
| Cell Cycle and Replication | |
| 83540 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction