missing translation for 'onlineSavingsMsg'
Learn More

Nujol (light oil), Thermo Scientific Chemicals

Product Code. 15414277
Change view
Click to view available options
Quantity:
50 mL
500 mL
2500 mL
Unit Size:
2500mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414277 50 mL 50mL
15424277 500 mL 500mL
15404277 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15414277 Supplier Thermo Scientific Alfa Aesar Supplier No. H31661.AD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatorischer Hinweis: Dieses Produkt unterliegt der Verbrauchsteuer auf Energieerzeugnisse gemäß der europäischen Richtlinie 2003/96 / EWG.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *

Specifications

Melting Point ∼-20°C
Density 0.838
Boiling Point >300°C
Flash Point 220°C (428°F)
Refractive Index 1.4613
Quantity 50 mL
Chemical Name or Material Nujol (light oil)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.