Learn More
Invitrogen™ NuMA Recombinant Rabbit Monoclonal Antibody (24GB1290)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557527
 View more versions of this product
Product Code. 30274939
  • 198.00 € / 20µL
Description
NuMA Recombinant Monoclonal Antibody for Western Blot, Flow
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NuMA
Recombinant Monoclonal
Unconjugated
NUMA1
6720401E04Rik; AA764025; AL022610; AU014979; centrophilin stabilizes mitotic spindle in mitotic cells; NMP22; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NUMA; NuMA protein; Numa1; SP-H antigen; structural nuclear protein
Rabbit
Affinity chromatography
RUO
101706, 308870, 4926
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1290
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
E9Q7G0, Q14980
NUMA1
A synthesized peptide derived from human NUMA1 (aa 2000-2100).
20 μL
Primary
Human, Mouse, Rat