missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUMB Monoclonal Antibody (447716)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561422

Product Code. 30534209

  • 379.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Protein NUMB homolog is an evolutionarily conserved protein that controls the differentiation of neuronal progenitor cells. Recent studies have revealed that NUMB functions in clathrin-dependent endocytosis by binding to the AP-2 complex. NUMB is also expressed in postmitotic neurons and plays a role in axonal growth. Neural cells expressing Numb isoforms with short phosphotyrosine-binding (SPTB) domain undergo extensive neurite outgrowth, an effect that can be blocked by voltage-gated Ca2+ channel (VGCC) inhibitor or by Ca2+ chelator.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NUMB
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P49757
Numb
E. coli-derived recombinant human Numb, Thr356-Leu592.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot, Immunocytochemistry
447716
Unconjugated
Numb
c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171
Mouse
Protein A/G
RUO
8650
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.