Learn More
Thermo Scientific™ Nunc™ 384-Well Microplate, white, TC surface, Pack of 10
Product Code. 10080681
Change view
Click to view available options
Unit Size:
Pack of 30
Product Code. unitSize
10080681 Pack of 30
1 options
Product Code. 10080681
Brand: Thermo Scientific™ 164610
Only null left
Add to basket
Edge
Add to basket
Improve background, autofluorescence and crosstalk with Nunc 384-well polystyrene white microplates.
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc 384-Well Polystyrene Black and White Microplates.
  • Color: White
  • Sterility: Sterile
  • Lid: With lid
White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid With Lid
Culture Area 0.084 cm2/well
Array 16 x 24
Packaging 10 Pk., 30 Cases
Product Line Nunc
Color White
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Case 30
No. per Pack 10