Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
BIOPHARMA
Product Code. 10567131
Change view
Click to view available options
No. of Wells:
96
No. per Pack:
1 Microplate
10 Microplates
No. per Case:
160 Microplates
50 Microplates
Unit Size:
Pack of 160
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells No. per Pack No. per Case Unit Size
10567131 96 10 Microplates 160 Microplates Pack of 160
10212811 96 1 Microplate 50 Microplates Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10567131 Supplier Thermo Scientific™ Supplier No. 161093
Stem Cell Research Savings
Use promo code 28032
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Color: Clear; Sterility: Sterile; Lid: With Lid
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid With Lid
Description 96F well plate,Nunclon Delta, clear, with lid, Sterile (Pack of 10, Case of 160)
Array 8 x 12
Packaging 10 Pk., 160 Cases
Product Line Nunc
Color Clear
Product Type Microplate
Skirt Style High Flange
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Flat Bottom (F-well)
No. per Case 160 Microplates
No. per Pack 10 Microplates
Quantity 160/Cs.