missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUP188 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548940
This item is not returnable.
View return policy
Description
NUP188 Polyclonal Antibody for Western Blot
May function as a component of the nuclear pore complex (NPC).
Specifications
| NUP188 | |
| Polyclonal | |
| Unconjugated | |
| NUP188 | |
| BC025526; hNup188; KIAA0169; mKIAA0169; nucleoporin 188; nucleoporin 188kDa; nucleoporin NUP188 homolog; NUP188; U89435 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23511 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5SRE5 | |
| NUP188 | |
| KLH conjugated synthetic peptide between 132-160 amino acids from the N-terminal region of human NU188. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction