missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62 Antibody (SR2155), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22380-50ul
This item is not returnable.
View return policy
Description
NUP62 Monoclonal antibody specifically detects NUP62 in Human samples. It is validated for Western Blot
Specifications
| NUP62 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR2155 | |
| Western Blot 1:500-1:2000 | |
| DKFZp547L134,62 kDa nucleoporin, FLJ20822, FLJ43869, IBSNp62, MGC841, nuclear pore glycoprotein p62, nucleoporin 62kD, nucleoporin 62kDa, SNDINucleoporin Nup62 | |
| A synthesized peptide derived from human NUP62 (Uniprot #: P37198) | |
| 50 μg | |
| Apoptosis, Cell Biology, Signal Transduction | |
| 23636 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction