missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUPL2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94275-0.02ml
This item is not returnable.
View return policy
Description
NUPL2 Polyclonal antibody specifically detects NUPL2 in Human samples. It is validated for Western Blot
Specifications
| NUPL2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CG1NUP42 homolog, H_RG271G13.9, hCG1, NLP_1, NLP-1, Nucleoporin hCG1, nucleoporin like 2, nucleoporin-like protein 1, nucleoporin-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human NUPL2 (NP_031368.1). MAICQFFLQGRCRFGDRCWNEHPGARGAGGGRQQPQQQPSGNNRRGWNTTSQRYSNVIQPSSFSKSTPWGGSRDQEKPYFSSFDSGASTNRKEGFGLSENPFASLSPDEQKDEKKLLEGIVKDMEVWESSGQWMFSVYSPVKKKPNISGFTDISPEELRLEYHNFLTSNNLQSYLNSVQRLINQWRNRVNELKSLNISTK | |
| 0.02 mL | |
| Signal Transduction | |
| 11097 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction