missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NUTM1 Monoclonal Antibody (2B6)
GREENER_CHOICE

Product Code. 30535161
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535161 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535161 Supplier Invitrogen™ Supplier No. MA562068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NUTM1 Monoclonal Antibody for Western Blot, IHC (P)

Plays a role in the regulation of proliferation. Regulates TERT expression by modulating SP1 binding to TERT promoter binding sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUTM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene NUTM1
Gene Accession No. Q86Y26, Q8BHP2
Gene Alias C15orf55; FAM22H; Nuclear protein in testis; NUT; NUT family member 1; NUT midline carcinoma family member 1; NUT midline carcinoma, family member 1; NUTM1
Gene Symbols NUTM1
Host Species Mouse
Immunogen Partial recombinant protein made to an internal portion of human NUT (between amino acids 400-700).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213765, 256646
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.