missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nutrient Agar

Product Code. 13255339
Change view
Click to view available options
Description:
Nutrient Agar
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
13255339 Nutrient Agar Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13255339 Supplier Thermo Scientific™ Supplier No. PO0155A
  • ONLINE EXCLUSIVE PRICE
    14.50 €
    14.11 € / Pack of 10
    Save 0.39 €
    2% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Cultivate and isolate fastidious pathogens and other microorganisms without affecting their hemolytic reactions with this general purpose medium.

Cultivate and isolate fastidious pathogens and other microorganisms with clearly visible hemolytic reactions with Thermo Scientific™ Oxoid™ Nutrient Agar, a formulation designed to provide especially nutritious media to permit maximum recovery of delicate microorganisms without affecting their hemolytic reactions. An improved version of Blood Agar Base, the medium can be used without the supplementation of blood to subculture microorganisms for maintenance purposes, or to check the purity of subcultures from isolation plates prior to further testing.

Use Nutrient Agar for the cultivation of a variety of organisms without interfering with their hemolytic reactions.

  • Permits the maximum recovery of delicate microorganisms without affecting hemolytic reactions.
  • No need for supplementation when maintaining subcultures or preparing further biochemical or serological tests.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifica

Color Straw
Content And Storage 2°C to 10°C
Description Nutrient Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.