missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nutrient Agar (Slope)

Product Code. 12817202
Change view
Click to view available options
Quantity:
200/Pk.
24/Pk.
Fill Weight:
10.0g + 0.3g
3.0g + 0.3g
Packaging Type:
Twenty four bijou bottles with screw cap closures in a box
Twenty four universal bottles with screw cap closures in a box.
Two hundred bijou bottles with screw cap closures in a box.
Unit Size:
Each
Pack of 200
Pack of 24
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Fill Weight Packaging Type Unit Size
12817202 24/Pk. 3.0g + 0.3g Twenty four bijou bottles with screw cap closures in a box Each
12773586 200/Pk. 3.0g + 0.3g Two hundred bijou bottles with screw cap closures in a box. Pack of 200
12786939 24/Pk. 10.0g + 0.3g Twenty four universal bottles with screw cap closures in a box. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12817202 Supplier Thermo Scientific™ Supplier No. BO0336B
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate non-fastidious microorganisms for testing food, milk and dairy products using these nutritious, ready prepared slopes.

Cultivate non-fastidious microorganisms for testing food, milk and dairy products with Thermo Scientific™ Oxoid™ Nutrient Broth Slopes. This non-selective, fluid medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared Slopes of Nutrient Broth.

  • Suitable for general cultivation of less fasidious microorganisms.
  • Convenient: the pre-filled slopes are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Content And Storage 2 to 25°C
Description Nutrient Agar (Slope)
Format Bijou bottles
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 3.0g + 0.3g
Packaging Type Twenty four bijou bottles with screw cap closures in a box
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.