missing translation for 'onlineSavingsMsg'
Learn More

NVP 231, MedChemExpress

Product Code. 16412077
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412077 50 mg 50mg
16446366 100 mg 100mg
16425928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16412077 Supplier Medchem Express Supplier No. HY13945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP 231 is a potent, specific, and reversible ceramide kinase (CerK) inhibitor(IC50=12 nM) that competitively inhibits binding of ceramide to CerK. NVP 231 induces cell apoptosis by increasing DNA fragmentation and caspase-3 and caspase-9 cleavage.

NVP 231 is a potent, specific, and reversible ceramide kinase (CerK) inhibitor(IC50=12 nM) that competitively inhibits binding of ceramide to CerK. NVP 231 induces cell apoptosis by increasing DNA fragmentation and caspase-3 and caspase-9 cleavage.

Specifications

Chemical Name or Material NVP 231
Physical Form Solid
CAS 362003-83-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C25H25N3O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 41 mg/mL (95.01 mM)
SMILES O=C(C1(C[C@@H](C2)C3)C[C@@H]3C[C@@H]2C1)NC4=CC=C5N=C(NC(C6=CC=CC=C6)=O)SC5=C4
Molecular Weight (g/mol) 431.55
Formula Weight 431.55
Percent Purity 98.91%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.