missing translation for 'onlineSavingsMsg'
Learn More

Melford Nystatin

Product Code. 30018628
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018628 10 g 10g
30018629 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018628 Supplier Melford Supplier No. N8202010.0
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nystatin is an antifungal polyene antibiotic that is effective against a wide range of fungi, including yeast and molds.

Nystatin is an antifungal polyene antibiotic that is effective against a wide range of fungi, including yeast and molds. It works by binding to the fungal cell membrane, disrupting its integrity and leading to leakage of cellular contents.

Nystatin is particularly useful in preventing and treating fungal contamination in cell cultures, making it a valuable tool in microbiological research. Researchers often use Nystatin to maintain aseptic conditions in cell culture experiments.

It is added to growth media to prevent the overgrowth of fungi that could potentially compromise the integrity of cell lines and experimental results. It is selective for fungi and has minimal toxicity to mammalian cells at effective antifungal concentrations.

Specifications

Chemical Name or Material Nystatin
CAS 1400-61-9
Quantity 10 g
Molecular Formula C47H75NO17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.