missing translation for 'onlineSavingsMsg'
Learn More

Melford Nystatin

Product Code. p-200087146
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 30018628

Brand: Melford N8202010.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nystatin is an antifungal polyene antibiotic that is effective against a wide range of fungi, including yeast and molds.

Nystatin is an antifungal polyene antibiotic that is effective against a wide range of fungi, including yeast and molds. It works by binding to the fungal cell membrane, disrupting its integrity and leading to leakage of cellular contents.

Nystatin is particularly useful in preventing and treating fungal contamination in cell cultures, making it a valuable tool in microbiological research. Researchers often use Nystatin to maintain aseptic conditions in cell culture experiments.

It is added to growth media to prevent the overgrowth of fungi that could potentially compromise the integrity of cell lines and experimental results. It is selective for fungi and has minimal toxicity to mammalian cells at effective antifungal concentrations.

Specifications

Chemical Name or Material Nystatin
CAS 1400-61-9
Quantity 10 g
Molecular Formula C47H75NO17
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.