missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nystatin, 85+%

Product Code. 15555156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15555156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15555156 Supplier Thermo Scientific Chemicals Supplier No. 455500010
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Nystatin, CAS # 1400-61-9, is a polyene compound that shows anti-bacterial and antifungal activities. It is also an ionophore that increases the membrane permeability to some ions.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Nystatin is a polyene compound that shows anti-bacterial and antifungal activities against Streptomyces noursei, Streptomyces aureus, and other Streptomyces species

• It is also an ionophore with the capacity to increase membrane permeability

• Nystatin is a channel-forming ionophore and exerts its activity via formation of a membrane-spanning pore in the fungal plasma membrane, thus altering membrane permeability, allowing the leakage of intracellular contents, and the subsequent disruption of electrochemical gradients essential for proper cell function

Applications

• In in vitro experiments, Nystatin is an antifungal that is both fungistatic and fungicidal against a wide variety of yeasts and yeast-like fungi

• It has a higher selectivity for fungal cells over mammalian cells since nystatin presents greater binding affinity with ergosterol, a key sterol found in fungal cell walls, as opposed to its mammalian counterpart, cholesterol

• This compound is not active against organisms that do not contain sterols in their cell membrane (for example, bacteria)

• It is used on human macrophage cell line U937, to study its effect on the production of interleukin-8 by silver nanoparticle-treated U937 cell line

• It has the ability to prevent cell culture contamination as well as induce interleukin-1 and -8, and tumor necrosis factor α secretion in TLR2-expressing THP1 cells

• It modulates lipid raft profile in mice macrophages

TRUSTED_SUSTAINABILITY

Specifications

Type Nystatin
Physical Form Powder
CAS 1400-61-9
Melting Point >160.0°C
Color Brown to Yellow
Quantity 1 g
Molecular Formula C47H75NO17
MDL Number MFCD25541651
Solubility Information Insoluble in water and alochol. Freely soluble in DMF and dimethylsulfoxide slightly soluble in methanol
InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
Molecular Weight (g/mol) 926.107
PubChem CID 133640190
Formula Weight 926.1
Percent Purity ≥85%
Chemical Name or Material Nystatin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.