missing translation for 'onlineSavingsMsg'
Learn More

Nystatin, MP Biomedicals™

Product Code. 17620907 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 kU
Unit Size:
500ku
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17620907 500 kU 500ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17620907 Supplier MP Biomedicals Supplier No. 02194534.5
  • ONLINE EXCLUSIVE PRICE
    38.75 €
    36.75 € / 500ku
    Save 2.00 €
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nystatin is an antifungal antibiotic obtained from Streptomyces noursei.

Nystatin A is closely related to Amphotericin B. Each is a macrocyclic lactone containing a ketal ring, an all-trans polyene system, and a mycosamine (3-amino-3-deoxyrhamnose) moiety. Nystatin probably acts by binding to sterols in the cell membrane of the fungus with a resultant change in membrane permeability allowing leakage of intracellular components. It is absorbed very sparingly following oral administration. Most of the orally administered nystatin is passed unchanged in the stool.

  • Nystatin acts by binding to ergosterols in fungal cell membranes, increasing permeability of the membrane, and creating nystatin/ergosterol based ion channels.
  • It also functions as a lipid raft-inhibiting reagent and as membrane associated cholesterol.
  • It is known to be a mixture, but the composition has not been completely elucidated.
  • Working concentration is 50mg/L for yeasts and fungi.

    Specifications

    Physical Form Powder
    Absorbance 290, 305, 320 ± 2nm (max (ethanol))
    CAS 1400-61-9
    pH 6 to 8 (3% aq. soln.)
    Melting Point >160°C (literature)
    Loss on Drying <8%
    Quantity 500 kU
    Molecular Formula C47H75NO17
    Validated Application Cell Culture
    Synonym Mycostatin; Fungicidin
    Solubility Information Very slightly soluble in water (effective as a suspension); slightly to sparingly soluble in alcohol, in methanol, in n-propyl alcohol, and in n-butyl alcohol; insoluble in chloroform, in ether, and in benzene.
    InChI Key VQOXZBDYSJBXMA-AWCIUYGFSA-N
    SMILES CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O
    IUPAC Name (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb
    Molecular Weight (g/mol) 926.107
    PubChem CID 133640190
    Formula Weight 926.11g/mol
    Grade Not Available
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.