missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Nystatin ReadyMade™ Solution

Product Code. 30044042
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044042 10 x 1 mL 10mL
30044040 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044042 Supplier TOKU-E™ Supplier No. N09010x1ML
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nystatin ReadyMade™ Solution is provided as a sterile-filtered solution of Nystatin in water.

  • CAS Number: 1400-61-9
  • Molecular Formula: C47H75NO17
  • Molecular Weight: 926.09 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid
  • Source: Streptomyces noursei and Streptomyces aureus
  • Spectrum: Nystatin primarily targets fungi, yeasts, and mold. It is active against Candida species, but, in vitro, some species such as C. tropicalis can develop resistance. It is either fungistatic or fungicidal depending on the concentration of Nystatin, the pH of the medium, and the nature of the infecting organism. It has a slightly narrower spectrum than Amphotericin B
  • Mechanism of Action: Nystatin binds ergosterol in the fungal cell membrane forming pores and increasing permeability. It is a channel-forming ionophore, meaning it forms a membrane-spanning pore in the fungal plasma membrane. The altered permeability is toxic to the fungi leading to growth inhibition or death. It has a similar mechanism to Amphotericin B. It is not active against bacteria since they do not contain sterols in their cell membrane

Specifications

CAS 1400-61-9
Chemical Name or Material Nystatin A1
Color Colorless
Content And Storage -20°C, Protect from light
Description Nystatin ReadyMade™ Solution 20,000 to 40,000 units/mol in water
Concentration 20,000 to 40,000 units/mL
Form Clear, colorless solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Water
For Use With (Application) Microbiology, Plant Biology, Eukaryotic Cell Culture Application
Product Line ReadyMade™ Solutions
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Streptomyces noursei and Streptomyces aureus
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.