missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicylic acid, 99%

Product Code. 11454114
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454114 500 g 500g
11458710 100 g 100g
11464114 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454114 Supplier Thermo Scientific Alfa Aesar Supplier No. A12488.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-78-2 | C9H8O4 | 180.16 g/mol

O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.

Solubility
Soluble in 100% ethanol (80mg/ml), DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS), pH 7.2 (2.7mg/ml).

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

Melting Point 133°C to 140°C
Density 1.4
Flash Point 250°C (482°F)
Quantity 500 g
Beilstein 779271
Merck Index 14,851
Solubility Information Soluble in 100% ethanol (80mg/ml),DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS),pH 7.2 (2.7mg/ml).
Formula Weight 180.15
Percent Purity 99%
Chemical Name or Material O-Acetylsalicylic acid
compliance-icons
Product Identifier
  • O-Acetylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Acetylsalicylic acid(Aspirin)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.