missing translation for 'onlineSavingsMsg'
Learn More

o-Anisidine, 98+%

Product Code. 10192890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10192890 1 L Glass bottle 1 L
10010690 5 mL Glass bottle 5mL
10102900 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10192890 Supplier Thermo Scientific Acros Supplier No. 104810010
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90-04-0 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-04-0
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00007688
InChI Key VMPITZXILSNTON-UHFFFAOYSA-N
Synonym o-anisidine, 2-anisidine, ortho-anisidine, 2-aminoanisole, o-methoxyaniline, o-aminoanisole, benzenamine, 2-methoxy, 2-methoxybenzenamine, o-methoxyphenylamine, o-anisylamine
PubChem CID 7000
ChEBI CHEBI:82288
IUPAC Name 2-methoxyaniline
SMILES COC1=CC=CC=C1N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 3.0°C to 6.0°C
Density 1.0920g/mL
Boiling Point 225.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NH2
Refractive Index 1.5730 to 1.5750
Quantity 1 L
Beilstein 13, 358
Merck Index 14, 667
Specific Gravity 1.092
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in dilute mineral acid, miscible with alcohol and ether, soluble in acetone and benzene
Formula Weight 123.15
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material o-Anisidine, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • o-Anisidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.